Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDC48B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335198.1 | NP_001325037.1 | 2 - 3 | 316 | 68 | |
NM_126415.6 | NP_178463.1 | 2 - 3 | 317 | 68 | ||
GenBank mRNA | AY062443.1 | - | 2 - 3 | 258 | 68 | |
BT008371.1 | - | 2 - 3 | 207 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |