Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126418.2 | NP_178466.1 | 1 - 2 | 538 | 102 | |
NM_179599.2 | NP_849930.1 | 1 - 2 | 577 | 102 | ||
NM_201682.3 | NP_973411.1 | 1 - 2 | 572 | 102 | ||
GenBank mRNA | AY063894.1 | - | 1 - 2 | 517 | 102 | |
AY096386.1 | - | 1 - 2 | 184 | 102 | ||
AY141229.1 | - | 1 - 2 | 184 | 102 | ||
U81369.1 | - | 1 - 2 | 538 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Arabidopsis thaliana Probe spans exons |