Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GEK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335203.1 | NP_001325274.1 | 7 - 8 | 869 | 111 | |
NM_126426.5 | NP_178474.2 | 7 - 8 | 805 | 111 | ||
GenBank mRNA | AB091252.1 | - | 7 - 8 | 704 | 111 | |
AK228782.1 | - | 7 - 8 | 706 | 111 | ||
BT022079.1 | - | 7 - 8 | 692 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |