Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPK31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_148230.3 | NP_680596.2 | 4 - 5 | 1091 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |