Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CARB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102730.2 | NP_564338.1 | 2 - 3 | 2243 | 152 | |
GenBank mRNA | AF367268.1 | - | 2 - 3 | 2145 | 152 | |
AY133548.1 | - | 2 - 3 | 2035 | 152 | ||
U40341.1 | - | 2 - 3 | 2041 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |