Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102773.3 | NP_564354.1 | 1 - 2 | 682 | 71 | |
GenBank mRNA | AY035092.1 | - | 1 - 2 | 316 | 71 | |
AY063056.1 | - | 1 - 2 | 272 | 71 | ||
KJ920369.1 | - | 1 - 2 | 272 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |