Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332900.1 | NP_001323337.1 | 9 - 10 | 1581 | 78 | |
NM_102781.3 | NP_174333.2 | 11 - 12 | 1881 | 78 | ||
NM_179400.1 | NP_849731.1 | 11 - 12 | 1850 | 78 | ||
NM_179401.2 | NP_849732.1 | 11 - 12 | 1881 | 78 | ||
GenBank mRNA | AK226602.1 | - | 11 - 12 | 1836 | 78 | |
AM113986.1 | - | 11 - 12 | 1851 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |