Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FEI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198193.1 | NP_001185122.1 | 6 - 7 | 800 | 84 | |
NM_001332962.1 | NP_001320669.1 | 5 - 6 | 1197 | 84 | ||
NM_102881.4 | NP_174427.3 | 6 - 7 | 782 | 84 | ||
GenBank mRNA | FJ708643.1 | - | 5 - 6 | 455 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |