Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP97A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102914.3 | NP_564384.1 | 6 - 7 | 1025 | 107 | |
GenBank mRNA | AY056446.1 | - | 6 - 7 | 1020 | 107 | |
AY058173.1 | - | 6 - 7 | 1025 | 107 | ||
AY142017.1 | - | 6 - 7 | 987 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |