Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102940.6 | NP_174486.1 | 2 - 3 | 1078 | 124 | |
GenBank mRNA | AY044335.1 | - | 2 - 3 | 609 | 124 | |
AY084576.1 | - | 2 - 3 | 718 | 124 | ||
AY128355.1 | - | 2 - 3 | 715 | 124 | ||
BT000019.1 | - | 2 - 3 | 609 | 124 | ||
X58149.1 | - | 687 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |