Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AOX1D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333005.1 | NP_001320824.1 | 1 - 2 | 532 | 92 | |
NM_102968.3 | NP_564395.1 | 1 - 2 | 532 | 92 | ||
GenBank mRNA | BT029014.1 | - | 1 - 2 | 411 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |