Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ESP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198198.1 | NP_001185127.1 | 20 - 21 | 1753 | 77 | |
NM_102984.5 | NP_174527.2 | 20 - 21 | 1778 | 77 | ||
GenBank mRNA | AY074318.1 | - | 20 - 21 | 1701 | 77 | |
AY133872.1 | - | 19 - 20 | 1555 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |