Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127084.3 | NP_179126.2 | 2 - 3 | 438 | 102 | |
GenBank mRNA | AF530477.1 | - | 2 - 3 | 279 | 102 | |
AK229578.1 | - | 2 - 3 | 337 | 102 | ||
BT025547.1 | - | 2 - 3 | 252 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |