Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SQN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127141.3 | NP_565381.1 | 5 - 6 | 1071 | 95 | |
GenBank mRNA | AY026065.1 | - | 5 - 6 | 1071 | 95 | |
BT028999.1 | - | 5 - 6 | 916 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |