Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AT2G16730 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127225.3 | NP_179264.2 | 9 - 10 | 1261 | 61 | |
GenBank mRNA | AJ270309.1 | - | 9 - 10 | 1082 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |