Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHT3;3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127282.3 | NP_179319.1 | 2 - 3 | 849 | 85 | |
GenBank mRNA | AK227300.1 | - | 2 - 3 | 847 | 85 | |
BT010199.1 | - | 2 - 3 | 490 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |