Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS35A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127333.5 | NP_179370.2 | 13 - 14 | 1633 | 102 | |
GenBank mRNA | BT008553.1 | - | 13 - 14 | 1511 | 102 | |
BT008664.1 | - | 13 - 14 | 1561 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |