Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS35A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127333.5 | NP_179370.2 | 19 - 20 | 2172 | 73 | |
GenBank mRNA | BT008553.1 | - | 19 - 20 | 2050 | 73 | |
BT008664.1 | - | 19 - 20 | 2100 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |