Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPK16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127343.2 | NP_179379.1 | 11 - 12 | 1709 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |