Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036446.2 | NP_001031523.1 | 2 - 3 | 992 | 138 | |
NM_001202794.1 | NP_001189723.1 | 4 - 5 | 683 | 138 | ||
NM_001336892.1 | NP_001318399.1 | 1 - 2 | 608 | 138 | ||
NM_001336893.1 | NP_001318400.1 | 4 - 5 | 1626 | 138 | ||
NM_001336895.1 | NP_001324806.1 | 3 - 4 | 1341 | 138 | ||
NM_129675.5 | NP_181643.1 | 3 - 4 | 673 | 138 | ||
GenBank mRNA | AF424577.1 | - | 3 - 4 | 653 | 138 | |
AY120719.1 | - | 3 - 4 | 653 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |