Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BGLU29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001125052.1 | NP_001118524.1 | 10 - 11 | 1207 | 105 | |
NM_180086.1 | NP_850417.1 | 10 - 11 | 1207 | 105 | ||
GenBank mRNA | AK118055.1 | - | 10 - 11 | 1207 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |