Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DL1D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_180087.1 | NP_850418.1 | 8 - 9 | 813 | 139 | |
NM_180088.3 | NP_850419.1 | 8 - 9 | 947 | 139 | ||
NM_180089.2 | NP_850420.1 | 9 - 10 | 1006 | 139 | ||
GenBank mRNA | AF488807.1 | - | 8 - 9 | 813 | 139 | |
AJ304843.1 | - | 9 - 10 | 998 | 139 | ||
AJ304844.1 | - | 8 - 9 | 947 | 139 | ||
BT046114.1 | - | 9 - 10 | 864 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |