Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRF9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337133.1 | NP_001324015.1 | 1 - 2 | 552 | 78 | |
NM_001337134.1 | NP_001324014.1 | 1 - 2 | 461 | 78 | ||
NM_180107.3 | NP_850438.2 | 2 - 3 | 454 | 78 | ||
GenBank mRNA | AB493593.1 | - | 1 - 2 | 224 | 78 | |
AY074647.1 | - | 1 - 2 | 228 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |