Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP76C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337143.1 | NP_001325389.1 | 3 - 4 | 845 | 107 | |
NM_180108.2 | NP_850439.1 | 2 - 3 | 1239 | 107 | ||
GenBank mRNA | AY078939.1 | - | 2 - 3 | 1071 | 107 | |
AY124874.1 | - | 2 - 3 | 917 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |