Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GBF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337182.1 | NP_001323457.1 | 6 - 7 | 613 | 93 | |
NM_001337183.1 | NP_001323456.1 | 6 - 7 | 613 | 93 | ||
NM_130190.3 | NP_182150.1 | 6 - 7 | 628 | 93 | ||
NM_180118.2 | NP_850449.1 | 6 - 7 | 628 | 93 | ||
GenBank mRNA | AF370307.1 | - | 6 - 7 | 568 | 93 | |
AY063093.1 | - | 5 - 6 | 335 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |