Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337538.1 | NP_001319470.1 | 4 - 5 | 893 | 62 | |
NM_111335.2 | NP_566233.1 | 5 - 6 | 714 | 62 | ||
NM_180183.3 | NP_850514.1 | 5 - 6 | 738 | 62 | ||
GenBank mRNA | AK117325.1 | - | 5 - 6 | 579 | 62 | |
BT024507.1 | - | 4 - 5 | 492 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |