Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337538.1 | NP_001319470.1 | 5 - 6 | 1002 | 57 | |
NM_111335.2 | NP_566233.1 | 6 - 7 | 823 | 57 | ||
NM_180183.3 | NP_850514.1 | 6 - 7 | 847 | 57 | ||
GenBank mRNA | AK117325.1 | - | 6 - 7 | 688 | 57 | |
BT024507.1 | - | 5 - 6 | 601 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |