Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337562.1 | NP_001325515.1 | 11 - 12 | 1576 | 106 | |
NM_111381.3 | NP_566246.1 | 11 - 12 | 1693 | 106 | ||
NM_180185.3 | NP_850516.1 | 11 - 12 | 1691 | 106 | ||
GenBank mRNA | AK226843.1 | - | 11 - 12 | 1651 | 106 | |
AY056292.1 | - | 11 - 12 | 1601 | 106 | ||
AY127022.1 | - | 11 - 12 | 1644 | 106 | ||
AY142674.1 | - | 10 - 11 | 1292 | 106 | ||
BT000631.1 | - | 10 - 11 | 1292 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |