Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATSK12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111458.5 | NP_187235.1 | 3 - 4 | 678 | 91 | |
NM_180189.4 | NP_850520.1 | 3 - 4 | 406 | 91 | ||
GenBank mRNA | AK227034.1 | - | 3 - 4 | 446 | 91 | |
AK316673.1 | - | 3 - 4 | 475 | 91 | ||
AY062713.1 | - | 3 - 4 | 391 | 91 | ||
AY093347.1 | - | 2 - 3 | 200 | 91 | ||
X75431.1 | - | 373 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |