Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WRKY13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120101.3 | NP_195651.1 | 2 - 3 | 835 | 112 | |
GenBank mRNA | AB493729.1 | - | 2 - 3 | 731 | 112 | |
AF421153.1 | - | 2 - 3 | 731 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Arabidopsis thaliana Probe spans exons |