Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP96A12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120111.3 | NP_195661.1 | 1 | 1015 | 97 | |
GenBank mRNA | AY058837.1 | - | 1 | 832 | 97 | |
BT003025.1 | - | 1 | 788 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |