Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204040.1 | NP_001190969.1 | 10 - 11 | 961 | 115 | |
NM_120114.2 | NP_195664.2 | 9 - 10 | 1063 | 115 | ||
NM_202986.2 | NP_974715.1 | 10 - 11 | 929 | 115 | ||
GenBank mRNA | AK117791.1 | - | 9 - 10 | 1063 | 115 | |
BT005291.1 | - | 9 - 10 | 836 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |