Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342547.1 | NP_001328757.1 | 8 - 9 | 1611 | 86 | |
NM_120126.5 | NP_568064.1 | 8 - 9 | 1556 | 86 | ||
GenBank mRNA | AK227118.1 | - | 8 - 9 | 1418 | 86 | |
AY054264.1 | - | 8 - 9 | 1410 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |