Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120162.5 | NP_195708.1 | 1 - 2 | 665 | 76 | |
GenBank mRNA | AY090349.1 | - | 1 - 2 | 595 | 76 | |
AY140052.1 | - | 1 - 2 | 595 | 76 | ||
BT000821.1 | - | 1 - 2 | 522 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |