Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120162.5 | NP_195708.1 | 4 - 5 | 1468 | 65 | |
GenBank mRNA | AY090349.1 | - | 4 - 5 | 1398 | 65 | |
AY140052.1 | - | 4 - 5 | 1398 | 65 | ||
BT000821.1 | - | 4 - 5 | 1325 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |