Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UNE18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342644.1 | NP_001330946.1 | 7 - 8 | 960 | 100 | |
NM_001342645.1 | NP_001330945.1 | 6 - 7 | 761 | 100 | ||
NM_120288.3 | NP_195830.1 | 7 - 8 | 1158 | 100 | ||
GenBank mRNA | AY050839.1 | - | 7 - 8 | 919 | 100 | |
AY091166.1 | - | 7 - 8 | 800 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |