Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCI20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203276.1 | NP_001190205.1 | 1 - 2 | 241 | 79 | |
NM_120305.3 | NP_195847.1 | 1 - 2 | 403 | 79 | ||
GenBank mRNA | AF370554.1 | - | 1 - 2 | 183 | 79 | |
BT008725.1 | - | 1 - 2 | 172 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |