Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCI20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120305.3 | NP_195847.1 | 3 - 4 | 663 | 80 | |
GenBank mRNA | AF370554.1 | - | 3 - 4 | 443 | 80 | |
AY086132.1 | - | 3 - 4 | 471 | 80 | ||
BT008725.1 | - | 3 - 4 | 432 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |