Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRR7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342671.1 | NP_001330923.1 | 4 - 5 | 1219 | 90 | |
NM_120359.3 | NP_568107.1 | 5 - 6 | 1082 | 90 | ||
GenBank mRNA | AB046954.1 | - | 4 - 5 | 772 | 90 | |
AY039943.1 | - | 5 - 6 | 1032 | 90 | ||
AY142560.1 | - | 4 - 5 | 772 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |