Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IREG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203288.1 | NP_001190217.1 | 6 - 7 | 1333 | 66 | |
NM_001342716.1 | NP_001332410.1 | 4 - 5 | 1579 | 66 | ||
NM_001342717.1 | NP_001332409.1 | 3 - 4 | 1050 | 66 | ||
NM_120438.4 | NP_195977.2 | 5 - 6 | 1503 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |