Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCB29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120473.3 | NP_196011.1 | 1 - 2 | 661 | 95 | |
GenBank mRNA | AY059727.1 | - | 1 - 2 | 656 | 95 | |
AY091365.1 | - | 1 - 2 | 594 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |