Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDH1-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127401.3 | NP_179435.1 | 7 - 8 | 920 | 141 | |
GenBank mRNA | AK119142.1 | - | 7 - 8 | 899 | 141 | |
BT005938.1 | - | 7 - 8 | 896 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |