Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHYB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335612.1 | NP_001325249.1 | 2 - 3 | 3291 | 144 | |
NM_127435.4 | NP_179469.1 | 2 - 3 | 3284 | 144 | ||
GenBank mRNA | X17342.1 | - | 3060 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |