Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHYB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127435.4 | NP_179469.1 | 3 - 4 | 3581 | 99 | |
GenBank mRNA | X17342.1 | - | 3357 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |