Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335621.1 | NP_001318247.1 | 3 - 4 | 610 | 67 | |
NM_001335622.1 | NP_001324425.1 | 4 - 5 | 962 | 67 | ||
NM_127468.5 | NP_179501.1 | 4 - 5 | 764 | 67 | ||
GenBank mRNA | AJ297264.1 | - | 3 - 4 | 569 | 67 | |
AK227102.1 | - | 4 - 5 | 673 | 67 | ||
AY096796.1 | - | 4 - 5 | 670 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |