Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EER5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335638.1 | NP_001324728.1 | 9 - 10 | 729 | 81 | |
NM_127514.4 | NP_179546.2 | 11 - 12 | 1081 | 81 | ||
GenBank mRNA | AK118897.1 | - | 11 - 12 | 1052 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |