Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NADP-ME1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335646.1 | NP_001324890.1 | 11 - 12 | 983 | 74 | |
NM_127548.4 | NP_179580.1 | 13 - 14 | 1356 | 74 | ||
GenBank mRNA | AY062734.1 | - | 13 - 14 | 1250 | 74 | |
BT003371.1 | - | 13 - 14 | 1200 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |