Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XIG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335672.1 | NP_001325368.1 | 26 - 27 | 3409 | 85 | |
NM_127588.3 | NP_179619.2 | 26 - 27 | 3218 | 85 |
Set Membership: |
![]() ![]() ![]() ![]() |