Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RALFL14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127630.1 | NP_179658.1 | 1 | 246 | 64 | |
GenBank mRNA | DQ056538.1 | - | 1 | 246 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |