Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pEARLI4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335728.1 | NP_001323931.1 | 2 - 3 | 302 | 92 | |
NM_001335729.1 | NP_001323932.1 | 2 - 3 | 302 | 92 | ||
NM_001335730.1 | NP_001323930.1 | 1 - 2 | 476 | 92 | ||
NM_127665.3 | NP_179692.1 | 2 - 3 | 270 | 92 | ||
GenBank mRNA | AY059813.1 | - | 2 - 3 | 264 | 92 | |
BT000010.1 | - | 1 - 2 | 130 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |